Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177572.1 | NP_001171043.1 | 7 | 808 | ||
NM_015829.3 | NP_056644.1 | 7 | 808 | |||
GenBank mRNA | AF164632.1 | - | 7 | 753 | ||
AK002829.1 | - | 7 | 683 | |||
AK012670.1 | - | 7 | 699 | |||
AK029020.1 | - | 7 | 739 | |||
AK150496.1 | - | 7 | 754 | |||
AK151180.1 | - | 7 | 754 | |||
AK152237.1 | - | 7 | 754 |