Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caprin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111289.1 | NP_001104759.1 | 14 | 1820 | ||
NM_001111290.1 | NP_001104760.1 | 14 | 1793 | |||
NM_001111291.1 | NP_001104761.1 | 15 | 1860 | |||
NM_001111292.1 | NP_001104762.1 | 14 | 1820 | |||
NM_016739.3 | NP_058019.2 | 15 | 1860 | |||
GenBank mRNA | AB373955.1 | - | 14 | 1682 | ||
AK133891.1 | - | 15 | 1823 | |||
AK141992.1 | - | 14 | 1762 | |||
AK147514.1 | - | 12 | 5902 | |||
AK149183.1 | - | 1 | 61 | |||
AK157707.1 | - | 15 | 1844 | |||
AK161131.1 | - | 9 | 955 | |||
AK166461.1 | - | 14 | 1778 | |||
AK168676.1 | - | 14 | 1788 | |||
AK169366.1 | - | 6 | 591 | |||
AK170830.1 | - | 15 | 1822 | |||
BC052427.1 | - | 15 | 1839 | 6836041 | ||
BC127030.1 | - | 6 | 686 | |||
U18773.1 | - | 4 | 481 | |||
U27838.1 | - | 14 | 1797 |