Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018797.2 | NP_061267.1 | 23, 24 | 3891 | ||
GenBank mRNA | AF190578.1 | - | 23, 24 | 3891 | ||
AK033998.1 | - | 5, 6 | 751 |