Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh18a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019698.2 | NP_062672.2 | 11 | 1374 | ||
NM_153554.2 | NP_705782.2 | 11 | 1368 | |||
GenBank mRNA | AF056573.1 | - | 11 | 1279 | ||
AF056574.1 | - | 11 | 1273 | |||
AK134148.1 | - | 11 | 1334 | |||
AK147711.1 | - | 11 | 1294 | |||
AK151072.1 | - | 11 | 1296 | |||
AK153946.1 | - | 11 | 1295 | |||
AK159610.1 | - | 11 | 1296 | |||
AK166459.1 | - | 11 | 1260 | |||
AK166494.1 | - | 11 | 1261 | |||
AK168905.1 | - | 11 | 1294 | |||
AK200973.1 | - | 2 | 111 | |||
AK202353.1 | - | 114 | ||||
BC033427.1 | - | 11 | 1305 | 5009115 | ||
BC037699.1 | - | 11 | 1253 | 4188900 |