Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020506.1 | NP_065252.1 | 2 | 158 | ||
GenBank mRNA | AF145021.1 | - | 2 | 158 | ||
AK021022.1 | - | 2 | 634 | |||
AK084907.1 | - | 2 | 188 | |||
AK129430.1 | - | 2 | 153 | |||
AK163516.1 | - | 2 | 439 | |||
AK163558.1 | - | 2 | 171 |
Gene Symbol: Lats2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029242.1 | - | 3045 |