Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199275.2 | NP_001186204.1 | 4 | 453 | ||
NM_001199276.2 | NP_001186205.1 | 2 | 123 | |||
NM_001271455.1 | NP_001258384.1 | 3 | 176 | |||
NM_001271456.1 | NP_001258385.1 | 4 | 348 | |||
NM_021327.4 | NP_067302.2 | 3 | 295 | |||
GenBank mRNA | AJ242777.1 | - | 2 | 95 | ||
AJ242778.1 | - | 3 | 267 | |||
AK135338.1 | - | 3 | 274 | |||
AK156131.1 | - | 3 | 147 | |||
AK170841.1 | - | 2 | 101 | |||
AK172890.1 | - | 81 | ||||
BC008186.1 | - | 3 | 212 | 3589311 | ||
BC008665.1 | - | 4 | 319 | 3598934 | ||
BY170310.1 | - | 2 | 122 | |||
BY197901.1 | - | 2 | 123 | |||
CJ175540.1 | - | 2 | 118 |