Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161818.1 | NP_001155290.1 | 7 | 822 | ||
NM_144519.4 | NP_653102.1 | 7 | 923 | |||
GenBank mRNA | AK049934.1 | - | 7 | 802 | ||
AK076145.1 | - | 7 | 924 | |||
AK076837.1 | - | 7 | 799 | |||
AK078000.1 | - | 7 | 823 | |||
AK201027.1 | - | 1 | 98 | |||
AK219010.1 | - | 1 | 19 | |||
AY736186.1 | - | 4 | 508 | |||
BC003941.1 | - | 5 | 572 | 3487151 | ||
BC042660.1 | - | 1 | 164 | |||
DQ323614.1 | - | 3 | 362 |