Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024209.2 | NP_077171.1 | 5, 6 | 561 | ||
GenBank mRNA | AK002764.1 | - | 5, 6 | 523 | ||
AK009104.1 | - | 5, 6 | 541 | |||
BC002223.1 | - | 5, 6 | 586 | 3491111 |