Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159632.1 | NP_001153104.1 | 14 | 1349 | ||
NM_133699.2 | NP_598460.1 | 13 | 1319 | |||
GenBank mRNA | AB088357.1 | - | 13 | 1304 | ||
AK004157.1 | - | 10 | 1033 | |||
BC056636.1 | - | 14 | 1322 | 5065583 | ||
BC064071.1 | - | 12 | 1150 |
Gene Symbol: Pdia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK148928.1 | - | 2933 |