Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178616.3 | NP_848731.2 | 13 | 1273 | ||
GenBank mRNA | AK007547.1 | - | 13 | 1242 | ||
AK012951.1 | - | 12 | 1737 | |||
AK029365.1 | - | 9 | 844 | |||
AK051738.1 | - | 13 | 1263 | |||
AK076002.1 | - | 13 | 1261 | |||
AK089942.1 | - | 13 | 1238 | |||
AK136049.1 | - | 13 | 1236 | |||
AK140773.1 | - | 1258 | ||||
AK167559.1 | - | 13 | 1244 | |||
AK168289.1 | - | 13 | 1265 | |||
AK204271.1 | - | 2 | 145 | |||
BC030432.1 | - | 10 | 927 | |||
BC037590.1 | - | 13 | 1222 | |||
BC055457.1 | - | 13 | 1205 | |||
BC090664.1 | - | 13 | 1225 | 30677672 | ||
BC090980.1 | - | 13 | 1240 | 30283128 | ||
BC100595.1 | - | 13 | 1235 | 6767671 | ||
BC119136.1 | - | 13 | 1244 | 8733885 | ||
BC119138.1 | - | 13 | 1244 | 8733887 |