Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stambp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024239.2 | NP_077201.1 | 3 | 349 | ||
GenBank mRNA | AB010123.1 | - | 3 | 296 | ||
AK017600.1 | - | 3 | 304 | |||
AK019907.1 | - | 3 | 299 | |||
AK046995.1 | - | 3 | 301 | |||
AK136961.1 | - | 3 | 900 | |||
AK139391.1 | - | 3 | 324 | |||
AK168810.1 | - | 3 | 322 | |||
AK220541.1 | - | 2 | 6318 | |||
BC003497.1 | - | 3 | 292 | 3153829 | ||
BC006939.1 | - | 3 | 347 | 2649718 | ||
BC025111.1 | - | 3 | 291 | 4194332 |