Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssfa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080558.4 | NP_542125.3 | 7 | 648 | ||
GenBank mRNA | AB093303.1 | - | 7 | 523 | ||
AB120565.1 | - | 7 | 533 | |||
AK037213.1 | - | 5 | 358 | |||
AK169543.1 | - | 7 | 649 | |||
BC079535.1 | - | 1 | 27 | |||
BC122519.1 | - | 7 | 534 | |||
BC141883.1 | - | 7 | 533 | 40097831 | ||
CF539296.1 | - | 7 | 464 |