Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam175a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172405.3 | NP_765993.1 | 2 | 129 | ||
GenBank mRNA | AK014420.1 | - | 2 | 155 | ||
AK042001.1 | - | 2 | 138 | |||
AK042339.1 | - | 2 | 138 | |||
AK051816.1 | - | 2 | 130 | |||
BC029845.1 | - | 2 | 134 | 4010744 |