Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Susd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162913.1 | NP_001156385.1 | 9, 10 | 1594 | ||
NM_027890.5 | NP_082166.3 | 11, 12 | 1954 | |||
GenBank mRNA | AK004703.2 | - | 11, 12 | 1924 | ||
AK028817.1 | - | 11, 12 | 1930 | |||
AK033823.1 | - | 9, 10 | 1567 | |||
AK037059.1 | - | 11, 12 | 1936 | |||
AK037127.1 | - | 11, 12 | 1905 | |||
AK038944.1 | - | 9, 10 | 2222 | |||
AK042469.1 | - | 11, 12 | 1938 | |||
AK128959.1 | - | 11, 12 | 1926 | |||
AK167155.1 | - | 10, 11 | 2136 | |||
BC026837.1 | - | 11, 12 | 1907 | 4222578 | ||
BC066833.1 | - | 8, 9 | 1753 |