Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sulf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252578.1 | NP_001239507.1 | 12 | 2078 | ||
NM_001252579.1 | NP_001239508.1 | 12 | 1924 | |||
NM_028072.5 | NP_082348.2 | 12 | 1920 | |||
GenBank mRNA | AK008108.1 | - | 3 | 183 | ||
AK028874.1 | - | 12 | 1835 | |||
AK034712.1 | - | 12 | 1905 | |||
AK036685.1 | - | 12 | 1856 | |||
AK049170.1 | - | 7 | 835 | |||
AK081643.1 | - | 12 | 1878 | |||
AK129316.1 | - | 12 | 2078 | |||
AK133336.1 | - | 1894 | ||||
AK165183.1 | - | 12 | 1832 | |||
AY101177.1 | - | 11 | 1739 | |||
BC062900.1 | - | 12 | 1876 | |||
BC141086.1 | - | 11 | 1673 | 9055698 |