Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164798.1 | NP_001158270.1 | 17 | 3386 | ||
NM_001164799.1 | NP_001158271.1 | 16 | 3365 | |||
NM_001164800.1 | NP_001158272.1 | 16 | 3362 | |||
NM_024249.5 | NP_077211.4 | 17 | 3389 | |||
GenBank mRNA | AK032245.1 | - | 17 | 3313 | ||
AK159948.1 | - | 16 | 3327 | |||
AK165672.1 | - | 16 | 3329 | |||
AK212080.1 | - | 1 | 44 | |||
AK216539.1 | - | 1 | 44 | |||
BC005641.1 | - | 5 | 1738 | |||
BC023375.1 | - | 1 | 518 | |||
BC043299.1 | - | 7 | 2062 | |||
BC055017.1 | - | 4 | 1410 | |||
BC082300.1 | - | 16 | 3333 | 30362204 | ||
BC088811.1 | - | 17 | 3322 | 6493546 |