Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ush1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163733.1 | NP_001157205.1 | 23 | 2366 | ||
NM_001291182.1 | NP_001278111.1 | 23 | 2366 | |||
NM_023649.2 | NP_076138.2 | 18 | 1454 | |||
NM_153677.2 | NP_710143.2 | 23 | 2366 | |||
GenBank mRNA | AF228924.1 | - | 18 | 1454 | ||
AF228925.1 | - | 23 | 2366 | |||
AK008274.2 | - | 2 | 123 | |||
AY103465.1 | - | 23 | 2366 | |||
AY246556.1 | - | 23 | 2340 | |||
BC010819.1 | - | 18 | 1456 | 4217086 | ||
BY708209.1 | - | 2 | 120 |