Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019588.2 | NP_062534.2 | 25 | 5765 | ||
GenBank mRNA | AB076247.1 | - | 25 | 5765 | ||
AF233885.1 | - | 4 | 725 | |||
AK122521.1 | - | 19 | 2951 | |||
BC138349.1 | - | 25 | 5753 | 8861369 | ||
BC138350.1 | - | 25 | 5753 | 8861370 | ||
BC145246.1 | - | 26 | 5795 | 9053798 |