Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nuak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028778.4 | NP_083054.1 | 2, 3 | 459 | ||
GenBank mRNA | AK004737.2 | - | 2, 3 | 447 | ||
AK033672.1 | - | 2, 3 | 446 | |||
BC033302.1 | - | 2, 3 | 434 | 3663595 |