Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029408.2 | NP_083684.1 | 3 | 932 | ||
GenBank mRNA | AK017032.1 | - | 3 | 932 | ||
AK076996.1 | - | 3 | 939 | |||
BC100441.1 | - | 3 | 921 | |||
BC125429.1 | - | 3 | 926 | 40129944 | ||
BC137644.1 | - | 3 | 926 | 8860663 |