Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029447.2 | NP_083723.1 | 5 | 737 | ||
GenBank mRNA | AK079090.1 | - | 5 | 738 | ||
AK138399.1 | - | 5 | 710 | |||
AK146364.1 | - | 5 | 686 | |||
AK170508.1 | - | 5 | 738 | |||
AK171432.1 | - | 5 | 738 | |||
BC016224.1 | - | 5 | 710 | 4486941 |