Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133761.3 | NP_598522.3 | 3 | 385 | ||
GenBank mRNA | AJ344447.1 | - | 3 | 385 | ||
AK047460.1 | - | 3 | 316 | |||
AK171707.1 | - | 3 | 309 | |||
BC066173.1 | - | 3 | 338 | 30049516 | ||
DV651307.1 | - | 3 | 415 |