Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030113.2 | NP_084389.2 | 7, 8 | 826 | ||
GenBank mRNA | AF297029.1 | - | 7, 8 | 730 | ||
AF297030.1 | - | 7, 8 | 789 | |||
AY179965.1 | - | 7, 8 | 702 | |||
AY179966.1 | - | 7, 8 | 702 | |||
AY179967.1 | - | 7, 8 | 702 | |||
CN703944.1 | - | 13 | ||||
CV783737.1 | - | 2, 3 | 204 |