Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242378.1 | NP_001229307.1 | 11 | 1367 | ||
NM_001242379.1 | NP_001229308.1 | 10 | 1277 | |||
NM_001242380.1 | NP_001229309.1 | 11 | 1367 | |||
NM_001242381.1 | NP_001229310.1 | 10 | 1274 | |||
NM_001242382.1 | NP_001229311.1 | 11 | 1364 | |||
NM_001242383.1 | NP_001229312.1 | 10 | 1274 | |||
NM_033552.3 | NP_291030.2 | 10 | 1277 | |||
GenBank mRNA | AB033759.1 | - | 10 | 1198 | ||
AK043017.1 | - | 10 | 1253 | |||
AK220501.1 | - | 11 | 1332 | |||
BC039226.1 | - | 10 | 1226 | 4480841 | ||
HM149278.1 | - | 10 | 1238 | |||
HQ533147.1 | - | 11 | 1304 | |||
HQ825316.1 | - | 11 | 1307 | |||
HQ831517.1 | - | 10 | 1214 | |||
JF500487.1 | - | 10 | 1354 | |||
JF500488.1 | - | 11 | 1444 | |||
JF500489.1 | - | 10 | 1354 | |||
JF500490.1 | - | 11 | 1444 | |||
JX220977.1 | - | 11 | 1331 | |||
JX254917.1 | - | 10 | 1244 | |||
JX268544.1 | - | 11 | 1445 |