Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242378.1 | NP_001229307.1 | 17 | 2298 | ||
NM_001242379.1 | NP_001229308.1 | 16 | 2208 | |||
NM_001242380.1 | NP_001229309.1 | 17 | 2298 | |||
NM_001242381.1 | NP_001229310.1 | 16 | 2205 | |||
NM_001242382.1 | NP_001229311.1 | 17 | 2295 | |||
NM_001242383.1 | NP_001229312.1 | 16 | 2205 | |||
NM_033552.3 | NP_291030.2 | 16 | 2208 | |||
GenBank mRNA | AB033759.1 | - | 16 | 2129 | ||
AK043017.1 | - | 16 | 2184 | |||
AK220501.1 | - | 17 | 2263 | |||
BC039226.1 | - | 16 | 2157 | 4480841 | ||
HM149278.1 | - | 16 | 2169 | |||
HQ533147.1 | - | 17 | 2235 | |||
HQ825316.1 | - | 17 | 2238 | |||
HQ831517.1 | - | 16 | 2145 | |||
JF500487.1 | - | 16 | 2285 | |||
JF500488.1 | - | 17 | 2375 | |||
JF500489.1 | - | 16 | 2285 | |||
JF500490.1 | - | 17 | 2375 | |||
JX220977.1 | - | 17 | 2262 | |||
JX254917.1 | - | 16 | 2175 | |||
JX268544.1 | - | 17 | 2376 |