Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007743.2 | NP_031769.2 | 11, 12 | 712 | ||
GenBank mRNA | AK132496.1 | - | 11, 12 | 708 | ||
AK159212.1 | - | 11, 12 | 713 | |||
AK159408.1 | - | 11, 12 | 709 | |||
AK159578.1 | - | 11, 12 | 709 | |||
AK160098.1 | - | 11, 12 | 712 | |||
AK160787.1 | - | 11, 12 | 708 | |||
AK160788.1 | - | 11, 12 | 707 | |||
AK160790.1 | - | 11, 12 | 708 | |||
AK160814.1 | - | 3, 4 | 177 | |||
AK160946.1 | - | 11, 12 | 711 | |||
AK161017.1 | - | 11, 12 | 713 | |||
AK161605.1 | - | 11, 12 | 708 | |||
AK164808.1 | - | 11, 12 | 708 | |||
BC007158.1 | - | 11, 12 | 692 | 3487477 | ||
BC042503.1 | - | 11, 12 | 700 | 3586272 | ||
X58251.1 | - | 11, 12 | 648 |