Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080466.2 | NP_536714.2 | 1 | 1190 | ||
GenBank mRNA | AF357241.1 | - | 1 | 930 | ||
AK137738.1 | - | 1 | 1191 | |||
AK170218.1 | - | 1 | 1193 | |||
BC106769.1 | - | 1 | 932 | 40039722 | ||
BC106770.1 | - | 1 | 932 | 40039723 | ||
DQ140280.1 | - | 1 | 882 |