Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nefm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008691.2 | NP_032717.2 | 3 | 1528 | ||
GenBank mRNA | AK018234.1 | - | 3 | 1367 | ||
AK051296.1 | - | 3 | 1371 | |||
AK051696.1 | - | 3 | 1368 | |||
AK080794.1 | - | 3 | 1372 | |||
AK164318.1 | - | 3 | 1371 | |||
AK165041.1 | - | 3 | 1371 | |||
BC052352.1 | - | 3 | 1370 | |||
BC058333.1 | - | 3 | 1361 | |||
BC119602.2 | - | 3 | 1336 | |||
BC128564.1 | - | 3 | 1331 | |||
DQ201636.1 | - | 3 | 1346 | |||
M20481.1 | - | 3 | 374 |