Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nefm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008691.2 | NP_032717.2 | 1 | 632 | ||
GenBank mRNA | AK018234.1 | - | 1 | 473 | ||
AK051296.1 | - | 1 | 475 | |||
AK051696.1 | - | 1 | 472 | |||
AK080794.1 | - | 1 | 476 | |||
AK164318.1 | - | 1 | 475 | |||
AK165041.1 | - | 1 | 475 | |||
BC052352.1 | - | 1 | 474 | |||
BC058333.1 | - | 1 | 465 | |||
BC119602.2 | - | 1 | 440 | |||
BC128564.1 | - | 1 | 435 | |||
DQ201636.1 | - | 1 | 450 |
Gene Symbol: A230070E04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK048964.1 | - | 1 | 619 | ||
AK162870.1 | - | 1 | 619 | |||
AK164995.1 | - | 1 | 686 |