Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291078.1 | NP_001278007.1 | 3 | 307 | ||
NM_011262.5 | NP_035392.1 | 3 | 307 | |||
GenBank mRNA | AK004812.1 | - | 3 | 244 | ||
AK138047.1 | - | 3 | 241 | |||
AK144106.1 | - | 3 | 246 | |||
AK144848.1 | - | 3 | 242 | |||
AK144954.1 | - | 3 | 253 | |||
AK145851.1 | - | 3 | 303 | |||
AK156252.1 | - | 2 | 77 | |||
AK178478.1 | - | 1 | 25 | |||
AK205804.1 | - | 2 | 106 | |||
BB839618.1 | - | 3 | 309 | |||
BC007188.1 | - | 2 | 95 | |||
BC012709.1 | - | 3 | 237 | 4208205 | ||
BY294588.1 | - | 3 | 300 | |||
CX207082.1 | - | 3 | 231 | |||
U10435.1 | - | 3 | 194 | |||
U43921.1 | - | 3 | 259 |