Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009105.4 | NP_033131.2 | 2 | 312 | ||
GenBank mRNA | AK009288.1 | - | 2 | 205 | ||
AK011857.1 | - | 2 | 205 | |||
AK135491.1 | - | 2 | 205 | |||
AK137622.1 | - | 2 | 208 | |||
AK149835.1 | - | 2 | 182 | |||
AK150011.1 | - | 2 | 180 | |||
AK152655.1 | - | 2 | 192 | |||
AK169076.1 | - | 2 | 230 | |||
AK172340.1 | - | 2 | 182 | |||
BC003827.1 | - | 2 | 175 | 3493122 | ||
CX228720.1 | - | 2 | 312 | |||
X63039.1 | - | 2 | 127 |