Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148938.3 | NP_683740.1 | 2 | 690 | ||
GenBank mRNA | AF330257.1 | - | 1 | 190 | ||
AK031831.1 | - | 2 | 350 | |||
AK040798.1 | - | 2 | 350 | |||
AK045716.1 | - | 2 | 300 | |||
AK045949.1 | - | 2 | 369 | |||
AK049090.1 | - | 2 | 350 | |||
AK080879.1 | - | 2 | 373 | |||
AK082736.1 | - | 2 | 350 | |||
AK084968.1 | - | 2 | 334 | |||
AK158777.1 | - | 1 | 224 | |||
AK162575.1 | - | 2 | 347 | |||
BC028638.1 | - | 2 | 292 | |||
BC058711.1 | - | 2 | 613 | 6842500 | ||
BC066154.1 | - | 2 | 293 | 30102325 |