Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcorl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163073.1 | NP_001156545.1 | 6 | 734 | ||
NM_172153.3 | NP_742165.1 | 6 | 747 | |||
NM_178142.5 | NP_835278.2 | 6 | 734 | |||
GenBank mRNA | AB076078.1 | - | 6 | 746 | ||
AK041987.1 | - | 6 | 693 | |||
AK042567.1 | - | 6 | 748 | |||
AK043498.1 | - | 6 | 692 | |||
AK043845.1 | - | 6 | 702 | |||
AK154398.1 | - | 6 | 695 | |||
AK155421.1 | - | 6 | 695 | |||
BC138563.1 | - | 6 | 734 | 8861583 |