Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177833.1 | NP_001171304.1 | 8 | 1929 | ||
NM_001177834.1 | NP_001171305.1 | 7 | 1750 | |||
NM_001177835.1 | NP_001171306.1 | 6 | 1678 | |||
NM_001177836.1 | NP_001171307.1 | 8 | 1431 | |||
NM_001177837.1 | NP_001171308.1 | 7 | 1329 | |||
NM_001177838.1 | NP_001171309.1 | 5 | 918 | |||
NM_001177839.1 | NP_001171310.1 | 6 | 1079 | |||
NM_001177840.1 | NP_001171311.1 | 4 | 744 | |||
NM_145533.2 | NP_663508.1 | 7 | 1839 | |||
GenBank mRNA | AF495851.1 | - | 6 | 947 | ||
AF495852.1 | - | 8 | 1300 | |||
AF495853.1 | - | 7 | 1708 | |||
AI466340.1 | - | 4 | 442 | |||
AJ567473.1 | - | 8 | 1848 | |||
AJ567474.1 | - | 6 | 1597 | |||
AJ567475.1 | - | 4 | 663 | |||
AJ567476.1 | - | 7 | 1669 | |||
AJ567477.1 | - | 6 | 998 | |||
AJ567478.1 | - | 5 | 837 | |||
AJ567479.1 | - | 7 | 1115 | |||
AJ567480.1 | - | 6 | 1901 | |||
AK143129.1 | - | 7 | 1813 | |||
AK159275.1 | - | 7 | 1766 | |||
AK159372.1 | - | 7 | 1766 | |||
AK166350.1 | - | 1228 | ||||
BC004831.1 | - | 7 | 1758 | 2647695 | ||
BQ174872.1 | - | 1 | 348 |