Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other VMN1R-PS79 SIRNA Assays ›
          Gene Symbol:
          Vmn1r-ps79 Gm10665 Gm10666 Gm10670 Gm4175 Gm4207 Gm4214 Vmn1r114 Vmn1r121 Vmn1r126 Vmn1r93
          Gene Name:
          vomeronasal 1 receptor, pseudogene 79
          predicted gene 10665
          predicted gene 10666
          predicted gene 10670
          predicted gene 4175
          vomeronasal 1 receptor Vmn1r-ps81 pseudogene
          predicted gene 4214
          vomeronasal 1 receptor 114
          vomeronasal 1 receptor 121
          vomeronasal 1 receptor 126
          vomeronasal 1 receptor 93
          Gene Aliases:
          Gm652
          ENSMUSG00000074308, Vmn1r150
          ENSMUSG00000074310, Vmn1r141
          ENSMUSG00000074330, Vmn1r102
          Vmn1r133
          Vmn1r-ps81
          Vmn1r161
          ENSMUSG00000074323, Gm10669
          EG667240, Gm8533
          Gm4170
          V1rd21
          Chromosome Location:
          Chr.7: 20356626 - 20358918 on Build GRCm38
          Chromosome Location:
          Chr.7: 22463110 - 22464075 on Build GRCm38
          Chromosome Location:
          Chr.7: 22133776 - 22134741 on Build GRCm38
          Chromosome Location:
          Chr.7: 20473960 - 20474925 on Build GRCm38
          Chromosome Location:
          Chr.7: 21760531 - 21761475 on Build GRCm38
          Chromosome Location:
          Chr.7: 22806346 - 22808474 on Build GRCm38
          Chromosome Location:
          Chr.7: 22983729 - 22984673 on Build GRCm38
          Chromosome Location:
          Chr.7: 20811221 - 20812186 on Build GRCm38
          Chromosome Location:
          Chr.7: 21097569 - 21098513 on Build GRCm38
          Chromosome Location:
          Chr.7: 21300502 - 21301467 on Build GRCm38
          Chromosome Location:
          Chr.7: 20138123 - 20139088 on Build GRCm38
          UniGene ID:
          Mm.431911
          Mm.484986
          Mm.484998
          Mm.389840
          Mm.484994
          Mm.431911
          Mm.482051
          Mm.480291
          Mm.480259
          Mm.480289
          Mm.377932
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID MSS243401
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Vmn1r-ps79
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC115870.1 - 1 1257 40062110
          BC119492.1 - 1 197
          Gene Symbol: Gm10665
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001167160.1 NP_001160632.1 1 198
          Gene Symbol: Gm10666
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001167573.1 NP_001161045.1 1 198
          Gene Symbol: Gm10670
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001167161.1 NP_001160633.1 1 198
          GenBank mRNA BC107204.1 - 1 197
          Gene Symbol: Gm4175
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001167165.1 NP_001160637.1 1 198
          Gene Symbol: Gm4207
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC107205.1 - 1 197
          BC119491.1 - 197
          Gene Symbol: Gm4214
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001167164.1 NP_001160636.1 1 198
          Gene Symbol: Vmn1r114
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166837.1 NP_001160309.1 1 198
          Gene Symbol: Vmn1r121
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166741.1 NP_001160213.1 1 198
          Gene Symbol: Vmn1r126
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166838.1 NP_001160310.1 1 198
          Gene Symbol: Vmn1r93
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_207547.2 NP_997430.2 1 198

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0