Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025375.2 | NP_001020546.1 | 4 | 350 | ||
NM_001025376.2 | NP_001020547.1 | 4 | 239 | |||
NM_023191.3 | NP_075680.1 | 4 | 244 | |||
GenBank mRNA | AF309554.1 | - | 4 | 202 | ||
AK010283.1 | - | 4 | 190 | |||
AK012342.1 | - | 4 | 256 | |||
AK013114.1 | - | 4 | 208 | |||
AK049527.1 | - | 4 | 208 | |||
AK077037.1 | - | 284 | ||||
AK142498.1 | - | 4 | 207 | |||
AK153862.1 | - | 4 | 277 | |||
AK167706.1 | - | 4 | 277 | |||
BC023026.1 | - | 4 | 189 | 5374890 | ||
BY165863.1 | - | 4 | 210 | |||
EF011623.1 | - | 4 | 187 |