Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2410164B09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK010824.1 | - | 2 | 465 |
Gene Symbol: Sp3os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI450369.1 | - | 1 | 218 | ||
AK005870.1 | - | 3 | 284 | |||
AK013593.1 | - | 3 | 243 | |||
BY152393.1 | - | 3 | 294 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_183265.1 | - | 284 | Chr. - - [-] on Build |