Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026791.2 | NP_081067.2 | 8 | 1183 | ||
GenBank mRNA | AK003747.1 | - | 3 | 247 | ||
AK082085.1 | - | 7 | 1672 | |||
AK156709.1 | - | 7 | 1333 | |||
AK203100.1 | - | 2 | 68 | |||
AK211947.1 | - | 2 | 68 | |||
BC043658.1 | - | 8 | 1144 | |||
BC053694.1 | - | 6 | 599 | |||
BC060082.1 | - | 5 | 414 | |||
BC094896.1 | - | 1 | 815 | |||
BC144773.1 | - | 8 | 1549 | |||
BC145661.1 | - | 8 | 1549 |