Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039081.2 | NP_001034170.1 | 6, 7 | 658 | ||
NM_001286221.1 | NP_001273150.1 | 8, 9 | 789 | |||
NM_001286222.1 | NP_001273151.1 | 8, 9 | 843 | |||
NM_001286223.1 | NP_001273152.1 | 6, 7 | 639 | |||
NM_027696.3 | NP_081972.2 | 8, 9 | 846 | |||
GenBank mRNA | AK016915.1 | - | 1 | 88 | ||
AK129405.1 | - | 8, 9 | 766 | |||
AK168693.1 | - | 6, 7 | 639 | |||
AY769435.1 | - | 7, 8 | 638 | |||
AY769436.1 | - | 6, 7 | 561 | |||
AY769437.1 | - | 6, 7 | 611 | |||
AY769438.1 | - | 8, 9 | 711 | |||
BC043930.1 | - | 8, 9 | 771 | |||
BC108273.1 | - | 6, 7 | 625 | 30080243 | ||
BC130227.1 | - | 6, 7 | 610 | 40141863 | ||
BQ562973.1 | - | 3, 4 | 236 | |||
CN538186.1 | - | 3, 4 | 259 | |||
CN701986.1 | - | 1 | 12 |