Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164248.1 | NP_001157720.1 | 3, 4 | 461 | ||
NM_001164249.1 | NP_001157721.1 | 3, 4 | 461 | |||
NM_001164250.1 | NP_001157722.1 | 3, 4 | 461 | |||
NM_001164251.1 | NP_001157723.1 | 3, 4 | 461 | |||
NM_001164252.1 | NP_001157724.1 | 2, 3 | 434 | |||
NM_001164253.1 | NP_001157725.1 | 2, 3 | 434 | |||
NM_001164254.1 | NP_001157726.1 | 2, 3 | 434 | |||
NM_001164255.1 | NP_001157727.1 | 3, 4 | 461 | |||
NM_001164256.1 | NP_001157728.1 | 2, 3 | 434 | |||
NM_024427.4 | NP_077745.2 | 3, 4 | 461 | |||
GenBank mRNA | AK002271.1 | - | 3, 4 | 462 | ||
AK003175.1 | - | 3, 4 | 462 | |||
AK032942.1 | - | 3, 4 | 510 | |||
AK077713.1 | - | 3, 4 | 462 | |||
AK168640.1 | - | 3, 4 | 458 | |||
AK168783.1 | - | 3, 4 | 458 | |||
AK169188.1 | - | 3, 4 | 462 | |||
BC026720.1 | - | 3, 4 | 435 | |||
BC131643.1 | - | 40 | ||||
BC132035.1 | - | 3, 4 | 420 | 40130312 | ||
BC132037.1 | - | 3, 4 | 420 | 40130314 | ||
BC145307.1 | - | 3, 4 | 420 | 9053861 | ||
M22479.1 | - | 3, 4 | 434 | |||
X64831.1 | - | 3, 4 | 416 |