Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009523.2 | NP_033549.1 | 5 | 745 | ||
GenBank mRNA | AK012727.1 | - | 5 | 867 | ||
BC101962.1 | - | 5 | 745 | 40044945 | ||
BC103560.1 | - | 5 | 745 | 40044943 | ||
BC103561.1 | - | 5 | 745 | 40044944 | ||
BC103562.1 | - | 5 | 745 | 40044946 | ||
M89797.1 | - | 5 | 745 |