Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Homer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164086.1 | NP_001157558.1 | 7 | 832 | ||
NM_001164087.1 | NP_001157559.1 | 7 | 848 | |||
NM_011983.2 | NP_036113.1 | 7 | 865 | |||
GenBank mRNA | AB017136.1 | - | 7 | 858 | ||
AF093259.1 | - | 7 | 737 | |||
AF093260.1 | - | 7 | 770 | |||
AK158673.1 | - | 934 | ||||
BC038314.1 | - | 3 | 506 | |||
DV657327.1 | - | 7 | 770 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK133275.1 | - | 1882 | Chr. - - [-] on Build |