Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2510039O18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029841.3 | NP_084117.2 | 2 | 1976 | ||
GenBank mRNA | AK011059.1 | - | 2 | 1976 | ||
AK052853.1 | - | 1 | 837 | |||
AK153761.1 | - | 2 | 1977 | |||
AK173332.1 | - | 3 | 1831 | |||
AK204562.1 | - | 1 | 82 | |||
AK216363.1 | - | 1 | 83 | |||
BC012878.1 | - | 2 | 1949 | 4190292 | ||
CX230455.1 | - | 1 | 692 |