Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsf1os2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037770.1 | - | 296 | |||
AK041721.1 | - | 267 | ||||
AK047013.1 | - | 342 |