Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167967.2 | NP_001161439.1 | 5 | 877 | ||
NM_001281974.1 | NP_001268903.1 | 6 | 989 | |||
NM_001281975.1 | NP_001268904.1 | 6 | 1039 | |||
NM_133971.3 | NP_598732.2 | 5 | 877 | |||
GenBank mRNA | AK014768.1 | - | 6 | 868 | ||
AK031808.1 | - | 5 | 875 | |||
AK033623.1 | - | 5 | 875 | |||
AK038156.1 | - | 5 | 875 | |||
AK039879.1 | - | 3 | 322 | |||
AK042500.1 | - | 5 | 875 | |||
AK050983.1 | - | 5 | 878 | |||
AK081754.1 | - | 6 | 1037 | |||
AK082875.1 | - | 5 | 874 | |||
AK084456.1 | - | 6 | 987 | |||
AK088371.1 | - | 5 | 827 | |||
AK211198.1 | - | 2 | 127 | |||
BC002198.1 | - | 5 | 809 | 3489133 |