Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177992.2 | NP_818773.1 | 10 | 1184 | ||
GenBank mRNA | AK031004.1 | - | 10 | 1186 | ||
AK075772.1 | - | 10 | 1142 | |||
BC003886.1 | - | 10 | 1143 | 3590411 | ||
BC024109.1 | - | 10 | 1140 | 5149353 | ||
BE956837.1 | - | 1 | 373 |
Gene Symbol: Tinf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028952.1 | - | 8 | 2550 | ||
AK169480.1 | - | 3 | 2540 |