Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164804.1 | NP_001158276.1 | 10, 11 | 1280 | ||
NM_178893.4 | NP_849224.2 | 11, 12 | 1505 | |||
GenBank mRNA | AK030097.1 | - | 11, 12 | 1508 | ||
AK030971.1 | - | 10, 11 | 1237 | |||
AK054394.1 | - | 10, 11 | 1281 | |||
AK151197.1 | - | 3, 4 | 297 | |||
AK152038.1 | - | 3, 4 | 297 | |||
AK152105.1 | - | 3, 4 | 297 | |||
AK152490.1 | - | 10, 11 | 1269 | |||
AK153134.1 | - | 10, 11 | 1269 | |||
AK154247.1 | - | 10, 11 | 1237 | |||
AK170184.1 | - | 10, 11 | 1234 | |||
BC026634.1 | - | 2, 3 | 215 | |||
BC031164.1 | - | 8, 9 | 927 | |||
BC048864.1 | - | 1, 2 | 19 | |||
BC138731.1 | - | 10, 11 | 1305 | 8861753 | ||
BC138732.1 | - | 10, 11 | 1305 | 8861754 |