Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038643.1 | NP_001033732.1 | 9 | 1891 | ||
NM_023908.2 | NP_076397.2 | 9 | 1891 | |||
GenBank mRNA | AF226324.1 | - | 9 | 1789 | ||
AK031280.1 | - | 9 | 1894 | |||
AK037938.1 | - | 9 | 1706 | |||
AK046945.1 | - | 8 | 1251 | |||
AK152488.1 | - | 9 | 1725 | |||
AK166266.1 | - | 9 | 1662 | |||
BC025059.1 | - | 9 | 1724 | 2598913 | ||
BC094464.1 | - | 9 | 1683 |