Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ivns1abp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039511.1 | NP_001034600.1 | 5, 6 | 865 | ||
NM_001039512.1 | NP_001034601.1 | 5, 6 | 865 | |||
NM_054102.2 | NP_473443.2 | 5, 6 | 865 | |||
GenBank mRNA | AB055737.1 | - | 5, 6 | 861 | ||
AB055738.1 | - | 5, 6 | 861 | |||
AK007291.1 | - | 5, 6 | 815 | |||
AK129229.1 | - | 5, 6 | 803 | |||
AK144619.1 | - | 5, 6 | 844 | |||
AK144835.1 | - | 5, 6 | 846 | |||
AK145071.1 | - | 5, 6 | 856 | |||
AK145845.1 | - | 5, 6 | 815 | |||
AK146001.1 | - | 5, 6 | 846 | |||
AK146329.1 | - | 2, 3 | 189 | |||
AK146494.1 | - | 5, 6 | 818 | |||
AK146645.1 | - | 5, 6 | 816 | |||
AK159254.1 | - | 5, 6 | 847 | |||
BC004092.1 | - | 4, 5 | 565 | 3593827 | ||
BC040250.1 | - | 5, 6 | 857 | |||
DQ914522.1 | - | 5, 6 | 817 | |||
DQ914523.1 | - | 5, 6 | 787 |