Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113391.2 | NP_001106862.1 | 2 | 295 | ||
NM_001113392.2 | NP_001106863.1 | 2 | 295 | |||
NM_001113393.2 | NP_001106864.2 | 2 | 295 | |||
NM_031162.4 | NP_112439.1 | 2 | 295 | |||
GenBank mRNA | AK017904.1 | - | 2 | 258 | ||
AK088087.1 | - | 2 | 223 | |||
AK138774.1 | - | 2 | 296 | |||
BC052824.1 | - | 2 | 216 | 30074812 | ||
J04967.1 | - | 2 | 287 | |||
L03353.1 | - | 2 | 211 | |||
M33158.1 | - | 2 | 285 | |||
S51932.1 | - | 2 | 211 | |||
U56258.1 | - | 2 | 211 | |||
X84237.1 | - | 2 | 211 |